Skip to Content
Merck
All Photos(1)

Documents

RAB1146

Sigma-Aldrich

Rat IGF-I ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Igf1(24482)

General description

This ELISA antibody pair detects rat IGF-I and mouse IGF-I. Other species are not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenzheng Xia et al.
International journal of molecular medicine, 41(2), 1127-1137 (2017-12-06)
Doxorubicin (DOXO), an anthracycline antibiotic, is a commonly used anticancer drug. Despite its widespread usage, the therapeutic effects of DOXO are limited by its cardiotoxicity. Mesenchymal stem cell (MSC)-based therapies have had positive outcomes in the treatment of DOXO-induced cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service