Skip to Content
Merck
All Photos(2)

Documents

R6256

Sigma-Aldrich

13-cis-Retinal

≥85% (HPLC)

Synonym(s):

13-cis-Vitamin A aldehyde, Vitamin A aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥85% (HPLC)

form

powder

color

yellow to yellow-orange

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H]C(=O)C(/[H])=C(C)\C=C\C=C(C)\C=C\C1=C(C)CCCC1(C)C

InChI

1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,15H,7,10,14H2,1-5H3/b9-6+,12-11+,16-8+,17-13-

InChI key

NCYCYZXNIZJOKI-HWCYFHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Stecher et al.
Biochemistry, 38(41), 13542-13550 (1999-10-16)
Photoisomerization of 11-cis-retinal to all-trans-retinal triggers phototransduction in the retinal photoreceptor cells and causes ultimately the sensation of vision. 11-cis-Retinal is enzymatically regenerated through a complex set of reactions in adjacent retinal pigment epithelial cells (RPE). In this study using
Valentyn I Prokhorenko et al.
Science (New York, N.Y.), 313(5791), 1257-1261 (2006-09-02)
Optical control of the primary step of photoisomerization of the retinal molecule in bacteriorhodopsin from the all-trans to the 13-cis state was demonstrated under weak field conditions (where only 1 of 300 retinal molecules absorbs a photon during the excitation
Oliver Grauer et al.
Journal of neuro-oncology, 104(3), 801-809 (2011-03-05)
The objective of this prospective, monocentric phase-II pilot study was to evaluate toxicity and efficacy of neoadjuvant temozolomide (TMZ) and 13-cis retinoic acid (13-cRA) treatment in patients with newly diagnosed anaplastic gliomas after total or subtotal tumor resection. The primary
J K McBee et al.
Biochemistry, 39(37), 11370-11380 (2000-09-14)
In the retinal rod and cone photoreceptors, light photoactivates rhodopsin or cone visual pigments by converting 11-cis-retinal to all-trans-retinal, the process that ultimately results in phototransduction and visual sensation. The production of 11-cis-retinal in adjacent retinal pigment epithelial (RPE) cells
Alexandre Santos et al.
Cancer research, 64(9), 3223-3229 (2004-05-06)
Topoisomerase I inhibitors, such as CPT-11, are potent anticancer drugs against neuroblastoma (NB). Differentiating agents, such as retinoids, improve the survival of children with metastatic NB. To characterize the biological effects associated with exposure to CPT-11 in vivo, athymic mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service