Skip to Content
Merck
All Photos(1)

Key Documents

EP0030125150

Eppendorf® Microtube 3810X

capacity 1.5 mL, colorless, pack of 1000 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.11

material

cap (plug seal)
colorless
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube

feature

graduations: no
neck thread Push fit

packaging

pack of 1000 ea

manufacturer/tradename

Eppendorf® 0030125150

capacity

1.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

The easy to open and close design of the Eppendorf Tubes® 3810X offers convenient, ergonomic and reliable one-hand operation. Whether sample preparation, centrifugation or storage - the Eppendorf Tubes® 3810X fit perfectly into your workflows!

Application

Eppendorf® Microtube 3810X is suitable for sample preparation, centrifugation, and storage.

Features and Benefits

  • Ensures convenient ergonomic operation with easy to open and close lid.
  • Polypropylene protects against chemical and mechanical stress and extreme temperature.
  • ["g-Safe"]: Provides great centrifugation stability and prevents sample spill due to tube damage and extended protection against hazardous samples.
  • Graduation for visual control of the pipetted volume
  • Frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Tube lid is easy to open and close for ergonomic operation
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes g-Safe®: Exceptional centrifugation stability up to 30,000 x g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • Graduation for visual control of the pipetted volume
  • Frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Other Notes

Manufactured without slip agents, plasticizers, and biocides substances that negatively affect bio-assay results.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service