Skip to Content
Merck
All Photos(1)

Key Documents

IM 9

86051302, human bone marrow, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

IM 9, 86051302

biological source

human bone marrow

growth mode

Suspension

karyotype

2n = 46

morphology

Lymphoblast

products

Immunoglobulin G (IgG)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human B lymphocyte

Cell Line Description

Derived from bone marrow removed from a female patient with multiple myeloma. The cells synthesise IgG at a rate of 2.4ug/1,000,000 cells/day and have receptors for insulin, calcitonin and human growth hormone. They also exhibit surface markers and receptor sites characteristic of B lymphocytes.

Application

Biosynthetic and surface marker studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-4x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service