Skip to Content
Merck
All Photos(1)

Key Documents

ATG004

Sigma-Aldrich

A375 NY−ESO−1 Antigen Panel Cell Lines

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.41

biological source

human skin (malignant melanoma)

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

General description

This product is a panel of three (3) genetically modified cell lines involving the NY-ESO-1 gene. The panel consists of a NY-ESO-1 knockout cell line (NY-ESO-1-/-) as well as a NY-ESO-1low and a NY-ESO-1high expression cell line. The NY-ESO-1-/- cell line was generated using CompoZr® Zinc Finger Nuclease technology, and the two NY-ESO-1 expression cell lines were generated using MISSION® lentivirus. The NY-ESO-1low and NY-ESO-1high cell lines express NY-ESO-1 at low and high levels relative to the NY-ESO-1-/- cell line (NY-ESO-1high > NY-ESO-1low > NY-ESO-1-/-) as measured via fluorescence-activated cell sorting (FACS). As a comparison, wild type A375 cells demonstrate NY-ESO-1medium expression as measured via FACS.

The STR profile of these cell lines match that of its parental cell line - ECACC catalog number 88113005.

Cell Line Description

Derived from a 54 year old female with malignant melanoma. The cell line produces rapidly growing amelanotic melanomas in anti-thymocyte serum treated NIH Swiss mice.

Features and Benefits

The A375 NY-ESO-1 antigen panel cell lines feature NY-ESO-1 expressed at varying levels (KO, low, and high). These cell lines enable researchers to screen and evaluate the efficacy and potency of NY-ESO-1-targeted immunotherapies in a biologically-relevant cellular research model.

Components

ATG004A-1VL: A375 NY-ESO-1 Knockout
ATG004B-1VL: A375 NY-ESO-1 Low
ATG004C-1VL: A375 NY-ESO-1 High

Quality

Tested for Mycoplasma, bacterial and fungal content, post-freeze viability, and short terminal repeat (STR) analysis for cell line identification.

Legal Information

CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service