Skip to Content
Merck
All Photos(1)

Documents

97218

Millipore

Malt Extract Agar, modified

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

form

powder

Quality Level

composition

agar, 15 g/L
dextrin, 2.75 g/L
maltose, 12.75 g/L
peptic digest of animal tissue, 0.78 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

selective by low pH for Aspergillus spp.
selective by low pH for Candida spp.
selective by low pH for Penicillium spp.
selective by low pH for Pichia spp.
selective by low pH for Saccharomyces spp.
selective by low pH for Zygosaccharomyces spp.
selective by low pH for molds (General Media)
selective by low pH for yeasts (General Media)

Application

for isolation, detection and enumeration of yeasts and molds

Preparation Note

Suspend 31.28 g of Malt Extract Agar, Modified in 1000 ml of distilled water and add 2.35 g of glycerol (Cat. No. 49767). Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. AVOID OVERHEATING! The low pH (4.7) of the medium may catalyze the hydrolysis of the agar during prolonged heating, resulting in a softer gel or failure of the agar to gel.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service