Skip to Content
Merck
All Photos(2)

Key Documents

34228

Supelco

Triclosan-methyl

PESTANAL®, analytical standard

Synonym(s):

2,4,4′-Trichloro-2′-methoxydiphenyl ether, 5-Chloro-2-(2,4-dichlorophenoxy)anisole, Methyl triclosan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Cl3O2
CAS Number:
Molecular Weight:
303.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

COc1cc(Cl)ccc1Oc2ccc(Cl)cc2Cl

InChI

1S/C13H9Cl3O2/c1-17-13-7-9(15)3-5-12(13)18-11-4-2-8(14)6-10(11)16/h2-7H,1H3

InChI key

NLYDHBBTVWMLFD-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triclosan-methyl has been used as antimicrobial agent in assay as positive controls in the isolation and identification of active antimicrobial compounds from E. linza algae extract, which was analysed using reversed phase high performance liquid chromatography method (RP-HPLC) and gas chromatography method (GC) coupled with mass spectroscopy, and further confirmed by nuclear magnetic resonance studies (NMR).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Antimicrobial activities of stearidonic and gamma-linolenic acids from the green seaweed Enteromorpha linza against several oral pathogenic bacteria
Park N-H, et al.
Botanical studies, 54, 39-39 (2013)
Feng-Jiao Peng et al.
Environmental science & technology, 52(15), 8390-8398 (2018-07-17)
Personal care products are widely used in our daily life in considerable quantities and discharged via the down-the-drain route to aquatic environments, resulting in potential risks to aquatic organisms. We investigated bioaccumulation and biotransformation of two widely used personal care
Marina G Pintado-Herrera et al.
Journal of chromatography. A, 1429, 107-118 (2016-01-10)
The main goal of this work was to develop, optimize and validate a multi-residue method for the simultaneous determination of 97 contaminants, including fragrances, UV filters, repellents, endocrine disruptors, biocides, polycyclic aromatic hydrocarbons (PAHs), polychlorinated biphenyls (PCBs), organophosphorus flame retardants
Jackeline Stoski et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 52(12), 1133-1140 (2017-08-02)
The main objective of this study is to propose a new sorbent phase based on a commercial epoxy resin, for use as an alternative sorbent in the stir bar sorptive extraction (SBSE) technique. The analytes triclosan (TCS) and methyl-triclosan (MTCS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service