Skip to Content
Merck
All Photos(1)

Key Documents

XX2604702

Millipore

2-way valve, 1/4 x 1/4in. NPTM

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel (Series 300)

Sterilization Compatibility

autoclavable compatible

I.D.

6.3 mm (0.25 in.)

input

sample type bottled water
sample type beer
sample type pharmaceutical(s)
sample type cosmetics

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

Can be used in conjunction with Millipore filtration systems

Application

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Bioburden Testing, Water Monitoring

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service