Skip to Content
Merck
All Photos(1)

Key Documents

12-343

Sigma-Aldrich

Acetyl-Histone H4 (Lys5) Peptide

Acetyl-Histone H4 (Lys5) Peptide primarily used in Histone Acetyltransferase Assays (HAT).

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.42

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (histone acetyltransferase (HAT))

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Quality

Routinely evaluated by Immunoblot Analysis as a standard for antibodies specific for acetylation of lysine 5 of histone H4.

Physical form

sterile deionized water

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The malate-aspartate NADH shuttle member Aralar1 determines glucose metabolic fate, mitochondrial activity, and insulin secretion in beta cells.
Blanca Rubi,Araceli del Arco,Clarissa Bartley,Jorgina Satrustegui,Pierre Maechler
The Journal of Biological Chemistry null
Mutagenic effects of niridazole in animal-mediated and in liquid suspension assays using Escherichia coli K-12 as an indicator.
S Knasmuller,A Szakmary
Mutation Research null
C A Mizzen et al.
Cell, 87(7), 1261-1270 (1996-12-27)
The transcription initiation factor TFIID is a multimeric protein complex composed of TATA box-binding protein (TBP) and many TBP-associated factors (TAF(II)s). TAF(II)s are important cofactors that mediate activated transcription by providing interaction sites for distinct activators. Here, we present evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service