Skip to Content
Merck
All Photos(1)

Documents

777072

Sigma-Aldrich

Phthalimidomethyl butyl trithiocarbonate

97%

Synonym(s):

Butyl TTC protected amine RAFT, Protected methylamine butyl trithiocarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15NO2S3
CAS Number:
Molecular Weight:
325.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

88-93 °C

storage temp.

2-8°C

SMILES string

CCCCSC(=S)SCN1C(=O)c2ccccc2C1=O

InChI

1S/C14H15NO2S3/c1-2-3-8-19-14(18)20-9-15-12(16)10-6-4-5-7-11(10)13(15)17/h4-7H,2-3,8-9H2,1H3

InChI key

OUCYVVFRLVYRSW-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; Chain Transfer Agent (CTA)
Reversible Addition Fragmentation Chain Transfer (RAFT) Polymerization

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service