Skip to Content
Merck
All Photos(2)

Key Documents

520217

Sigma-Aldrich

Zirconium(IV) carbonate hydroxide oxide

Synonym(s):

Zirconium basic carbonate, Zirconium(IV) carbonate basic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zr(OH)2CO3·ZrO2
CAS Number:
Molecular Weight:
308.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
Pricing and availability is not currently available.

form

powder

Quality Level

mp

135 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O[Zr]1(O)OC(=O)O[Zr]2(O1)O[Zr](O)(O)O2

InChI

1S/CH2O3.4H2O.3O.3Zr/c2-1(3)4;;;;;;;;;;/h(H2,2,3,4);4*1H2;;;;;;/q;;;;;;;;+1;+2;+3/p-6

InChI key

YGCRTSJWNHPGDG-UHFFFAOYSA-H

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Does this become moist when standing in ambient conditions (50% humidity/ 23 degrees c)?

    1 answer
    1. The SDS for this product states that dry storage is recommended. The material is not specifically listed as being hygroscopic. While this item should be stored according to the SDS, it is not likely to rapidly uptake water under the conditions listed.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service