Merck
Control of Reversible Activation Dynamics of [Ru{η
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-07-25)
Francisco Martínez-Peña, Ana M Pizarro
ABSTRACT
The potential use of organoruthenium complexes as anticancer drugs is well known. Herein, a family of activatable tethered ruthenium(II) arene complexes of general formula [Ru{η
Sigma-Aldrich
2-Aminobiphenyl, ≥96.5% (GC)
Sigma-Aldrich
N,N′-Di-tert-butylethylenediamine, 98%
Sigma-Aldrich
N,N′-Diethylethylenediamine, 95%
Sigma-Aldrich
1,3-Diaminopropane, ≥99%