Merck
  • Dual Influence of Endocannabinoids on Long-Term Potentiation of Synaptic Transmission.
Dual Influence of Endocannabinoids on Long-Term Potentiation of Synaptic Transmission.
Frontiers in pharmacology (2018-01-10)
Armando Silva-Cruz, Mattias Carlström, Joaquim A Ribeiro, Ana M Sebastião
ABSTRACT
Cannabinoid receptor 1 (CB
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
JZL195, ≥98% (HPLC)
Sigma-Aldrich
AM251, >98% (HPLC), solid
Sigma-Aldrich
URB597, ≥98% (HPLC), powder