Merck
  • Comparative Study of the Adsorption of Thiols and Selenols on Au(111) and Au(100).
Comparative Study of the Adsorption of Thiols and Selenols on Au(111) and Au(100).
Langmuir : the ACS journal of surfaces and colloids (2017-11-08)
Nicolás Arisnabarreta, Gustavo D Ruano, Magalí Lingenfelder, E Martín Patrito, Fernando P Cometto
ABSTRACT
The effect of the Au crystalline plane on the adsorption of different thiols and selenols is studied via reductive desorption (RD) and X-ray photoelectron spectroscopy (XPS) measurements. Self-assembled monolayers (SAMs) using aliphatic (ATs) and aromatic thiols (ArTs) on both Au(111) and Au(100) were prepared. The electrochemical stability of these SAMs on both surfaces is evaluated by comparing the position of the RD peaks. The longer the AT chain the more stable the SAM on Au(100) when compared to Au(111). By means of XPS measurements, we determine that the binding energy (BE) of the S 2p signal corresponding to the S atoms at the thiol/Au interface, commonly assigned at 162.0 eV, shifts 0.2 eV from Au(111) to Au(100) for SAMs prepared using thiols with the C* (C atom bonded to S) in sp
Sigma-Aldrich
2-Mercaptopyridine-3-carboxylic acid, technical grade
Sigma-Aldrich
4-Methylbenzenethiol, 98%