Merck
  • Determination of Doxorubicin in Stealth Hyalurionic Acid-Based Nanoparticles in Rat Plasma by the Liquid-Liquid Nanoparticles-Breaking Extraction Method: Application to a Pharmacokinetic Study.
Determination of Doxorubicin in Stealth Hyalurionic Acid-Based Nanoparticles in Rat Plasma by the Liquid-Liquid Nanoparticles-Breaking Extraction Method: Application to a Pharmacokinetic Study.
Journal of chromatographic science (2016-06-01)
Xiaopeng Han, Wei Wei, Lu Zhong, Cong Luo, Chunnuan Wu, Qikun Jiang, Jin Sun
ABSTRACT
An efficient extraction of doxorubicin (Dox) from homemade stealth hyalurionic acid (HA)-based nanoparticles (NPs) in rat plasma could not be performed by previously published methods. Therefore, we attempted to establish the novel NPs-breaking and UPLC-MS-MS method for evaluating the pharmacokinetic profiles of the homemade stealth HA NPs in rats. The pretreatment method of plasma samples used the liquid-liquid extraction method with isopropyl alcohol as NPs-breaking and protein-precipitating solvents, and the NPs-breaking efficiency of isopropyl alcohol was as high as 97.2%. The analyte and gliclazide (internal standard) were extracted from plasma samples with isopropyl alcohol and were separated on UPLC BEH C18 with a mobile phase consisting of methanol and water (containing 0.1% formic acid). The method demonstrated good linearity at the concentrations ranging from 5 to 5,000 ng/mL. The intra- and interday relative standard deviations were >10%. Finally, the method was successfully applied to a pharmacokinetic study of homemade stealth HA-based NPs in rats following intravenous administration.
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%