Merck
  • A one-stage cultivation process for the production of poly-3-(hydroxybutyrate-co-hydroxyvalerate) from olive mill wastewater by Haloferax mediterranei.
A one-stage cultivation process for the production of poly-3-(hydroxybutyrate-co-hydroxyvalerate) from olive mill wastewater by Haloferax mediterranei.
New biotechnology (2016-05-26)
Diya Alsafadi, Othman Al-Mashaqbeh
ABSTRACT
Olive mill wastewater (OMW), a highly polluting waste from the olive oil industry, was utilized as sole carbon source for the production of polyhydroxyalkanoate (PHA) by extremely halophilic Haloferax Mediterranei (H. mediterranei) in a one stage cultivation step. H. mediterranei showed remarkable cell growth and tolerated the inhibitory effect of polyphenols present in medium containing 25% of OMW. H. mediterranei cultivation conditions were optimized in medium containing 15% OMW by investigating several parameters that affect the production of PHA. The highest polymer yield (0.2g/L) and PHA content (43% PHA/cell dry mass) were achieved at 37°C, 170rpm and 22% salt concentration. Analysis of the produced PHA revealed the production of copolyester poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBHV) containing 6.5mol% 3-hydroxyvalerate (3HV). The production of PHBHV was observed without the need for fermentation step or adding external carbon source. The PHBHV displayed reduced melting points at 140.1°C and 154.4°C when compared to homopolymer polyhydroxybutyrate.
Sigma-Aldrich
Methyl (R)-3-hydroxybutyrate, 99%
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
(−)-Methyl (R)-3-hydroxyvalerate, ≥98.0% (sum of enantiomers, GC)