Merck
  • DNase X is a glycosylphosphatidylinositol-anchored membrane enzyme that provides a barrier to endocytosis-mediated transfer of a foreign gene.
DNase X is a glycosylphosphatidylinositol-anchored membrane enzyme that provides a barrier to endocytosis-mediated transfer of a foreign gene.
The Journal of biological chemistry (2007-04-10)
Daisuke Shiokawa, Tokiyoshi Matsushita, Yukari Shika, Mamoru Shimizu, Masahiro Maeda, Sei-ichi Tanuma
ABSTRACT
DNase X is the first mammalian DNase to be isolated that is homologous to DNase I. In this study, we have examined its function using a novel monoclonal antibody and showed it to be expressed on the cell surface as a glycosylphosphatidylinositolanchored membrane protein. High level expression was observed in human muscular tissues and in myotubes obtained in vitro from RD rhabdomyosarcoma cells. We observed that RD myotubes incorporated a foreign gene, lacZ, by endocytosis but that expression of the encoded coding product, beta-galactosidase, was strongly inhibited. Overexpression of DNase X inhibited endocytosis-mediated gene transfer, whereas knockdown of DNase X with small interfering RNA had the opposite effect. These results reveal that DNase X provides a cell surface barrier to endocytosis-mediated gene transfer.
Sigma-Aldrich
Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Pan Cadherin antibody produced in rabbit, whole antiserum