Merck
Roscovitine targets, protein kinases and pyridoxal kinase.
The Journal of biological chemistry (2005-06-25)
Stéphane Bach, Marie Knockaert, Jens Reinhardt, Olivier Lozach, Sophie Schmitt, Blandine Baratte, Marcel Koken, Stephen P Coburn, Lin Tang, Tao Jiang, Dong-Cai Liang, Hervé Galons, Jean-Francois Dierick, Lorenzo A Pinna, Flavio Meggio, Frank Totzke, Christoph Schächtele, Andrea S Lerman, Amancio Carnero, Yongqin Wan, Nathanael Gray, Laurent Meijer
ABSTRACT
(R)-Roscovitine (CYC202) is often referred to as a "selective inhibitor of cyclin-dependent kinases." Besides its use as a biological tool in cell cycle, neuronal functions, and apoptosis studies, it is currently evaluated as a potential drug to treat cancers, neurodegenerative diseases, viral infections, and glomerulonephritis. We have investigated the selectivity of (R)-roscovitine using three different methods: 1) testing on a wide panel of purified kinases that, along with previously published data, now reaches 151 kinases; 2) identifying roscovitine-binding proteins from various tissue and cell types following their affinity chromatography purification on immobilized roscovitine; 3) investigating the effects of roscovitine on cells deprived of one of its targets, CDK2. Altogether, the results show that (R)-roscovitine is rather selective for CDKs, in fact most kinases are not affected. However, it binds an unexpected, non-protein kinase target, pyridoxal kinase, the enzyme responsible for phosphorylation and activation of vitamin B6. These results could help in interpreting the cellular actions of (R)-roscovitine but also in guiding the synthesis of more selective roscovitine analogs.
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Actin (Ab-1) Mouse mAb (JLA20), liquid, clone JLA20, Calbiochem®