Merck
  • The mitochondrial Na+/Ca2+ exchanger upregulates glucose dependent Ca2+ signalling linked to insulin secretion.
The mitochondrial Na+/Ca2+ exchanger upregulates glucose dependent Ca2+ signalling linked to insulin secretion.
PloS one (2012-10-12)
Iulia I Nita, Michal Hershfinkel, Daniel Fishman, Eyal Ozeri, Guy A Rutter, Stefano L Sensi, Daniel Khananshvili, Eli C Lewis, Israel Sekler
ABSTRACT
Mitochondria mediate dual metabolic and Ca(2+) shuttling activities. While the former is required for Ca(2+) signalling linked to insulin secretion, the role of the latter in β cell function has not been well understood, primarily because the molecular identity of the mitochondrial Ca(2+) transporters were elusive and the selectivity of their inhibitors was questionable. This study focuses on NCLX, the recently discovered mitochondrial Na(+)/Ca(2+) exchanger that is linked to Ca(2+) signalling in MIN6 and primary β cells. Suppression either of NCLX expression, using a siRNA construct (siNCLX) or of its activity, by a dominant negative construct (dnNCLX), enhanced mitochondrial Ca(2+) influx and blocked efflux induced by glucose or by cell depolarization. In addition, NCLX regulated basal, but not glucose-dependent changes, in metabolic rate, mitochondrial membrane potential and mitochondrial resting Ca(2+). Importantly, NCLX controlled the rate and amplitude of cytosolic Ca(2+) changes induced by depolarization or high glucose, indicating that NCLX is a critical and rate limiting component in the cross talk between mitochondrial and plasma membrane Ca(2+) signalling. Finally, knockdown of NCLX expression was followed by a delay in glucose-dependent insulin secretion. These findings suggest that the mitochondrial Na(+)/Ca(2+) exchanger, NCLX, shapes glucose-dependent mitochondrial and cytosolic Ca(2+) signals thereby regulating the temporal pattern of insulin secretion in β cells.
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis