Merck
  • Pharmacokinetics of intravitreally injected bevacizumab in vitrectomized eyes.
Pharmacokinetics of intravitreally injected bevacizumab in vitrectomized eyes.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2013-06-06)
Jeeyun Ahn, Hyuncheol Kim, Se Joon Woo, Ji Hyun Park, Sunyoung Park, Duck Jin Hwang, Kyu Hyung Park
ABSTRACT
To compare the pharmacokinetics (PKs) of intravitreally injected bevacizumab in vitrectomized versus nonvitrectomized control rabbit eyes. Twenty-five-gauge pars plana vitrectomy without lensectomy was performed in 17 right rabbit eyes (V) and 18 nonvitrectomized right rabbit eyes served as controls (C). After 1.25 mg/0.05 mL intravitreal bevacizumab (IVB) injections, eyes were enucleated at 1 h, 1, 2, 5, 14, and 30 days after the injection and immediately frozen at -80°C. Bevacizumab concentrations were determined after separation of frozen vitreous and aqueous humor (AH) compartments using indirect enzyme-linked immunosorbent assay. Bevacizumab concentration-time data were analyzed to obtain PK data. Vitreous clearance of IVB consisted of 2 phases, the first fast distribution and second slow elimination phase. Clearance of IVB was accelerated in V eyes only during the first phase and not in the second phase. The vitreous concentration percent ratios between V and C eyes were 94.7% (1 h), 70.5% (1 day), 89.2% (2 days), 94.2% (5 days), 99.2% (14 days), and 79.1% (30 days). Overall vitreous half-lives were 6.99 and 7.06 days for V and C eyes, respectively (1.6-h difference). Overall IVB PKs in rabbit eyes after vitrectomy without lensectomy are not substantially different from nonvitrectomized control eyes.
Sigma-Aldrich
Tropicamide, solid
Tropicamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture