Merck
  • Synthesis, characterization, biodegradation, and drug delivery application of biodegradable lactic/glycolic acid oligomers: I. Synthesis and characterization.
Synthesis, characterization, biodegradation, and drug delivery application of biodegradable lactic/glycolic acid oligomers: I. Synthesis and characterization.
Journal of biomaterials science. Polymer edition (1997-01-01)
N Wang, X S Wu, H Lujan-Upton, E Donahue, A Siddiqui
ABSTRACT
A series of oligomers or low molecular weight polymers of lactic and/or glycolic acid has been synthesized with different molar ratios of lactic to glycolic acid. These oligomers have been characterized with respect to oligomer composition, molecular weight, intrinsic viscosity, crystallinity, melting temperature, and glass transition temperature. The polymerization conditions for the lactic/glycolic acid oligomer syntheses were as follows: 180-220 degrees C, 5 mm Hg, 5 h, and 0.1 wt% of catalyst (antimony oxide) concentration. The polymeric compositions correlated to the feed ratios of lactic to glycolic acid. The molecular weight of the oligomers ranged from 895.8 +/- 48.7 to 1368.0 +/- 0 D with the intrinsic viscosity ranging from 0.0513 to 0.0814 dl g-1. The lactic/glycolic acid oligomers were found to be amorphous. The glass transition temperatures of the lactic/glycolic acid oligomers were lower than physiological temperature.
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O