- Comparative evaluation of NCCLS M27-A and EUCAST broth microdilution procedures for antifungal susceptibility testing of candida species.
Comparative evaluation of NCCLS M27-A and EUCAST broth microdilution procedures for antifungal susceptibility testing of candida species.
Antimicrobial agents and chemotherapy (2002-10-18)
Manuel Cuenca-Estrella, Wendy Lee-Yang, Meral A Ciblak, Beth A Arthington-Skaggs, Emilia Mellado, David W Warnock, Juan L Rodriguez-Tudela
PMID12384382
ABSTRACT
A two-laboratory study was performed to evaluate the correlation between the NCCLS M27-A and EUCAST microdilution procedures for antifungal testing of Candida spp. A panel of 109 bloodstream isolates was tested against amphotericin B, flucytosine, fluconazole, and itraconazole. Overall, the agreement was 92% and the intraclass correlation coefficient was 0.90 (P < 0.05).
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture