Merck
  • Piroxicam-beta-cyclodextrin. A review of its pharmacodynamic and pharmacokinetic properties, and therapeutic potential in rheumatic diseases and pain states.
Piroxicam-beta-cyclodextrin. A review of its pharmacodynamic and pharmacokinetic properties, and therapeutic potential in rheumatic diseases and pain states.
Drugs (1994-12-01)
C R Lee, J A Balfour
ABSTRACT
Piroxicam-beta-cyclodextrin is a complex of the established nonsteroidal antiinflammatory drug (NSAID) piroxicam and an inert cyclic macromolecule, beta-cyclodextrin. In clinical trials in patients with rheumatic diseases or pain arising from other conditions, it was as effective an analgesic as standard piroxicam, and showed a faster onset of action on the first day of treatment. In short term pharmacodynamic studies in healthy volunteers, piroxicam-beta-cyclodextrin was equivalent to or tended to show less gastrointestinal mucosal toxicity than standard piroxicam, as assessed by endoscopy and faecal blood loss. However, no data are available on its comparative gastrointestinal mucosal effects from long term clinical trials using similar measures. Preliminary findings from a clinical study suggest piroxicam-beta-cyclodextrin caused fewer gastroduodenal lesions than tenoxicam. As with other NSAIDs, the majority of adverse events associated with piroxicam-beta-cyclodextrin in clinical trials were gastrointestinal in origin, with epigastric pain, heartburn and nausea the most common. Thus, piroxicam-beta-cyclodextrin is an effective agent in patients with rheumatic diseases or other pain states. When rapid analgesia is required in the initial treatment of acute pain, the faster onset of action of piroxicam-beta-cyclodextrin may be an advantage over the parent compound; however, this is unlikely to be important during long term therapy. The results of further long term trials are awaited before firm conclusions can be reached regarding the gastrointestinal tolerability of piroxicam-beta-cyclodextrin compared with that of standard piroxicam and other NSAIDs.
Sigma-Aldrich
Piroxicam, ≥98% (TLC)
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Piroxicam, European Pharmacopoeia (EP) Reference Standard
USP
Piroxicam, United States Pharmacopeia (USP) Reference Standard
Piroxicam for system suitability, European Pharmacopoeia (EP) Reference Standard