Merck
Spironolactone for hirsutism in polycystic ovary syndrome.
The Annals of pharmacotherapy (2005-08-04)
Natalie A Christy, Andrea S Franks, L Brian Cross
ABSTRACT
To evaluate the level of evidence for treatment of hirsutism associated with polycystic ovary syndrome (PCOS) with spironolactone. Studies and reports were located in MEDLINE (1966-January 2005), EMBASE, and International Pharmaceutical Abstracts through the second week of January 2005. Hirsutism is a common clinical problem and is often associated with PCOS. Research has been performed to assess the efficacy of spironolactone therapy in patients with hirsute characteristics. Five studies that evaluated the effectiveness of spironolactone for hirsutism in PCOS were identified and reviewed. The effects of multiple treatment options on the subjective and objective measures of hirsutism have displayed varying results. The outcomes reported to date have shown a positive trend toward using spironolactone in women with PCOS and hirsutism.
Sigma-Aldrich
Spironolactone, 97.0-103.0%
Spironolactone, European Pharmacopoeia (EP) Reference Standard
Spironolactone for system suitability, European Pharmacopoeia (EP) Reference Standard