Merck
  • β-Carotene-9',10'-oxygenase status modulates the impact of dietary tomato and lycopene on hepatic nuclear receptor-, stress-, and metabolism-related gene expression in mice.
β-Carotene-9',10'-oxygenase status modulates the impact of dietary tomato and lycopene on hepatic nuclear receptor-, stress-, and metabolism-related gene expression in mice.
The Journal of nutrition (2014-02-21)
Hsueh-Li Tan, Nancy E Moran, Morgan J Cichon, Ken M Riedl, Steven J Schwartz, John W Erdman, Dennis K Pearl, Jennifer M Thomas-Ahner, Steven K Clinton
ABSTRACT
Tomato and lycopene (ψ,ψ-carotene) consumption is hypothesized to protect against nonalcoholic steatohepatitis and hepatocarcinogenesis, processes that may depend upon diet and gene interactions. To investigate the interaction of tomato or lycopene feeding with β-carotene-9',10'-monooxygenase (Bco2) on hepatic metabolic and signaling pathways, male wild-type (WT) and Bco2(-/-) mice (3-wk-old; n = 36) were fed semi-purified control, 10% tomato powder-containing, or 0.25% lycopene beadlet-containing diets for 3 wk. Serum lycopene concentrations were higher in lycopene- and tomato-fed Bco2(-/-) mice compared with WT (P = 0.03). Tomato- and lycopene-fed mice had detectable hepatic apolipoprotein (apo)-6'-, apo-8'-, and apo-12'-lycopenal concentrations. Hepatic expression of β-carotene-15,15'-monooxygenase was increased in Bco2(-/-) mice compared with WT (P = 0.02), but not affected by diet. Evaluation of hepatic gene expression by focused quantitative reverse transcriptase-polymerase chain reaction arrays for nuclear receptors and coregulators (84 genes) and stress and metabolism (82 genes) genes indicates that tomato feeding affected 31 genes (≥1.5-fold, P < 0.05) and lycopene feeding affected 19 genes, 16 of which were affected by both diets. Lycopene down-regulation of 7 nuclear receptors and coregulators, estrogen-related receptor-α, histone deacetylase 3, nuclear receptor coactivator 4, RevErbA-β, glucocorticoid receptor, peroxisome proliferator-activated receptor (PPAR)-α, and PPAR-γ, coactivator 1 β was dependent upon interaction with Bco2 status. Lycopene and tomato feeding induced gene expression patterns consistent with decreased lipid uptake, decreased cell proliferation and mitosis, down-regulated aryl hydrocarbon receptor signaling, and decreased expression of genes involved in retinoid X receptor heterodimer activation. Tomato feeding also caused expression changes consistent with down-regulation of DNA synthesis and terpenoid metabolism. These data suggest tomato components, particularly lycopene, affect hepatic gene expression, potentially affecting hepatic responses to metabolic, infectious, or chemical stress.
Product Description
Supelco
Lycopene, analytical standard