Merck
  • Design and synthesis of indole-based peptoids as potent noncompetitive antagonists of transient receptor potential vanilloid 1.
Design and synthesis of indole-based peptoids as potent noncompetitive antagonists of transient receptor potential vanilloid 1.
Journal of medicinal chemistry (2007-11-08)
Martina Quintanar-Audelo, Asia Fernández-Carvajal, Wim Van Den Nest, Cristina Carreño, Antonio Ferrer-Montiel, Fernando Albericio
ABSTRACT
The vanilloid receptor subunit 1, or transient receptor potential vanilloid 1 (TRPV1), integrates physical and chemical stimuli in the peripheral nervous system, playing a key role in inflammatory pain. Identification of potent TRPV1 antagonists is thus an important goal of current neuropharmacology. Herein, we describe the solid-phase synthesis of a series of indole-based peptoids (N-alkylglycines) and the biological activity of the peptoids as novel TRPV1 antagonists. The potency and selectivity of the compounds were determined by electrophysiological recordings in Xenopus oocytes. The most potent and selective noncompetitive TRPV1 antagonist of the series, compound 7, represents an interesting pharmacophoric structure for analgesic lead optimization.
Sigma-Aldrich
2,4-Dichlorobenzaldehyde, 99%