Merck
  • Specific targeting of acetylcholinesterase and butyrylcholinesterase recognition sites. Rational design of novel, selective, and highly potent cholinesterase inhibitors.
Specific targeting of acetylcholinesterase and butyrylcholinesterase recognition sites. Rational design of novel, selective, and highly potent cholinesterase inhibitors.
Journal of medicinal chemistry (2002-12-28)
Luisa Savini, Alessandra Gaeta, Caterina Fattorusso, Bruno Catalanotti, Giuseppe Campiani, Luisa Chiasserini, Cesare Pellerano, Ettore Novellino, Dawn McKissic, Ashima Saxena
ABSTRACT
Tacrine-based AChE and BuChE inhibitors were designed by investigating the topology of the active site gorge of the two enzymes. The homobivalent ligands characterized by a nitrogen-bridged atom at the tether level could be considered among the most potent and selective cholinesterase inhibitors described to date. The nitrogen-containing homobivalent ligands 3e,g and the sulfur-containing 3h validated the hypothesis of extra sites of interaction in the AChE and BuChE active site gorges.
Sigma-Aldrich
Butyrylcholinesterase from equine serum, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Butyrylcholinesterase from equine serum, lyophilized powder, ≥900 units/mg protein