Merck
  • Distribution, behavior and fate of azole antifungals during mechanical, biological, and chemical treatments in sewage treatment plants in China.
Distribution, behavior and fate of azole antifungals during mechanical, biological, and chemical treatments in sewage treatment plants in China.
The Science of the total environment (2012-04-24)
Xianzhi Peng, Qiuxin Huang, Kun Zhang, Yiyi Yu, Zhifang Wang, Chunwei Wang
ABSTRACT
Residue of azole antifungals in the environment is of concern due to the environmental risks and persistence. Distribution, behavior, and fate of frequently used azole antifungal pharmaceuticals were investigated in wastewater at two sewage treatment plants (STPs) in China. Fluconazole, clotrimazole, econazole, ketoconazole, and miconazole were constantly detected at 1-1834 ng L(-1) in the wastewater. The latter four were also ubiquitously detected in sewage sludge. Fluconazole passed through treatment in the STPs and largely remained in the final effluent. On the contrary, biotransformation and sorption to sludge occurred to the other azoles. Ketoconazole was more readily bio-transformed, whereas clotrimazole, econazole, and miconazole were more likely to be adsorbed onto and persisted in sewage sludge. Lipophilicity plays the governing role on adsorption. The highest concentrations in the raw wastewater were observed in winter for the azole pharmaceuticals except for fluconazole. The seasonal difference was smoothed out after treatment in the STPs.
Product Description
Econazole nitrate, European Pharmacopoeia (EP) Reference Standard
Econazole, European Pharmacopoeia (EP) Reference Standard