Merck
  • Novel squalene-hopene cyclase inhibitors derived from hydroxycoumarins and hydroxyacetophenones.
Novel squalene-hopene cyclase inhibitors derived from hydroxycoumarins and hydroxyacetophenones.
Chemical & pharmaceutical bulletin (2004-10-07)
Giancarlo Cravotto, Gianni Balliano, Silvia Tagliapietra, Simonetta Oliaro-Bosso, Gian Mario Nano
ABSTRACT
Squalene-hopene cyclase (SHC) is a useful model enzyme for predicting molecular interactions with oxidosqualene cyclase (OSC). Structure--activity relationships were investigated for numerous coumarin-derived inhibitors of SHC, and structural simplifications are suggested. Both umbelliferone and 2,4-dihydroxyacetophenone provide convenient starting nuclei for the design of SHC inhibitors. Derivatives bearing an omega-epoxyfarnesyl moiety or just a plain alkyl chain showed an inhibitory effect on a recombinant SHC from Alicyclobacillus acidocaldarius expressed in Escherichia coli.
Product Description
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5%
Supelco
2′,6′-Dihydroxyacetophenone, matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
2′,6′-Dihydroxyacetophenone, 97%