Merck
Kinetic model for erythrocyte aggregation.
Blood cells, molecules & diseases (2000-02-08)
S M Bertoluzzo, A Bollini, M Rasia, A Raynal
ABSTRACT
It is well known that light transmission through blood is the most widely utilized method for the study of erythrocyte aggregation. The curves obtained had been considered empirically as exponential functions. In consequence, the process becomes characterized by an only parameter that varies with all the process factors without discrimination. In the present paper a mathematical model for RBC aggregation process is deduced in accordance with von Smoluchowski's theory about the kinetics of colloidal particles agglomeration. The equation fitted the experimental pattern of the RBC suspension optical transmittance closely and contained two parameters that estimate the most important characteristics of the aggregation process separately, i.e., (1) average size of rouleaux at equilibrium and (2) aggregation rate. The evaluation of the method was assessed by some factors affecting erythrocyte aggregation, such as temperature, plasma dilutions, Dextran 500, Dextran 70 and PVP 360, at different media concentrations, cellular membrane alteration by the alkylating agent TCEA, and decrease of medium osmolarity. Results were interpreted considering the process characteristics estimated by the parameters, and there were also compared with similar studies carried out by other authors with other methods. This analysis allowed us to conclude that the equation proposed is reliable and useful to study erythrocyte aggregation.
Sigma-Aldrich
Tris(2-chloroethyl)amine hydrochloride, 98%