Merck
  • Conservation of honey bee (Apis mellifera) sperm phospholipids during storage in the bee queen--a TLC/MALDI-TOF MS study.
Conservation of honey bee (Apis mellifera) sperm phospholipids during storage in the bee queen--a TLC/MALDI-TOF MS study.
Experimental gerontology (2013-01-03)
Jakob Wegener, Kristin Zschörnig, Kristin Onischke, Beate Fuchs, Jürgen Schiller, Karin Müller
ABSTRACT
The honey bee (Apis mellifera) is characterized by a high degree of phenotypic plasticity of senescence-related processes, and has therefore become a model organism of gerontological research. Sperm of honey bee drones can remain fertile for several years within the storage organ of queens. The reason for this longevity is unknown, but the suppression of lipid peroxidation seems to play a decisive role. Here, we examined the questions of whether spermatheca- and in vitro-stored honey bee sperm are indeed resistant to lipid peroxidation, and whether the nature of sperm lipids could explain this resistance. The lipid composition of bee sperm was determined by matrix-assisted laser desorption and ionization time-of-flight (MALDI-TOF) mass spectrometry (MS) combined with thin-layer chromatography (TLC). The positive ion mass spectra of drone sperm lipids are dominated by two glycerophosphocholine (GPC) species, although small amounts of sphingomyelins (SM) and glycerophosphoethanolamines (GPE) are also detectable after TLC. Alkyl/acyl and alkenyl/acyl compounds of GPC, and alkyl/acyl as well as diacyl compounds of GPE were detected containing oleyl, oleoyl, palmityl and palmitoyl as the most abundant residues. Assignments of all compounds have been additionally verified by enzymatic digestion and exposition to HCl. During incubation of sperm in the presence of air, characteristic lipid oxidation products such as lysophosphatidylcholine (LPC) appear. Inside the spermatheca, however, sperm lipids are obviously protected from oxidation and their composition does not change, even if they are stored over years. Our data support the view that the membrane composition of honey bee sperm could help to explain the extraordinary longevity of these cells.
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution