Merck
  • Para-aminosalicylic acid acts as an alternative substrate of folate metabolism in Mycobacterium tuberculosis.
Para-aminosalicylic acid acts as an alternative substrate of folate metabolism in Mycobacterium tuberculosis.
Science (New York, N.Y.) (2012-11-03)
Sumit Chakraborty, Todd Gruber, Clifton E Barry, Helena I Boshoff, Kyu Y Rhee
ABSTRACT
Folate biosynthesis is an established anti-infective target, and the antifolate para-aminosalicylic acid (PAS) was one of the first anti-infectives introduced into clinical practice on the basis of target-based drug discovery. Fifty years later, PAS continues to be used to treat tuberculosis. PAS is assumed to inhibit dihydropteroate synthase (DHPS) in Mycobacterium tuberculosis by mimicking the substrate p-aminobenzoate (PABA). However, we found that sulfonamide inhibitors of DHPS inhibited growth of M. tuberculosis only weakly because of their intracellular metabolism. In contrast, PAS served as a replacement substrate for DHPS. Products of PAS metabolism at this and subsequent steps in folate metabolism inhibited those enzymes, competing with their substrates. PAS is thus a prodrug that blocks growth of M. tuberculosis when its active forms are generated by enzymes in the pathway they poison.
Sigma-Aldrich
Sodium 4-aminosalicylate dihydrate, ≥97.0% (T)
Sigma-Aldrich
4-Aminosalicylic acid, 99%