Merck
  • Simultaneous determination of L-ascorbic acid, dopamine and uric acid with gold nanoparticles-β-cyclodextrin-graphene-modified electrode by square wave voltammetry.
Simultaneous determination of L-ascorbic acid, dopamine and uric acid with gold nanoparticles-β-cyclodextrin-graphene-modified electrode by square wave voltammetry.
Talanta (2012-04-10)
Xianqing Tian, Changming Cheng, Hongyan Yuan, Juan Du, Dan Xiao, Shunping Xie, Martin M F Choi
ABSTRACT
Graphene decorated with gold nanoparticles (AuNPs-β-CD-Gra) has been synthesized by in situ thermal reduction of graphene oxide and HAuCl(4) with β-cyclodextrin (β-CD) under alkaline condition. The AuNPs-β-CD-Gra product was well characterized by infrared spectroscopy, X-ray powder diffraction, scanning electron microscopy, high-resolution transmission electron microscopy, and selected area electron diffraction. This material was used to fabricate an AuNPs-β-CD-Gra-modified glassy carbon electrode (GCE) which showed excellent electro-oxidation of l-ascorbic acid (AA), dopamine (DA) and uric acid (UA) in 0.10 M NaH(2)PO(4)-HCl buffer solution (pH 2.0) by square wave voltammetry (SWV). Three well-resolved oxidation peaks of AA and DA and UA were obtained. The AuNPs-β-CD-Gra/GCE exhibits linear responses to AA, DA and UA in the ranges 30-2000, 0.5-150 and 0.5-60 μM, respectively. The detection limits (based on S/N=3 and preconcentration time=3.0 min) for AA, DA and UA are 10, 0.15 and 0.21 μM, respectively. The AuNPs-β-CD-Gra/GCE has been successfully applied to determine UA in human urine with satisfactory results. Our work provides a simple, convenient and green route to synthesize AuNPs on Gra which is potentially useful in electroanalysis.
Sigma-Aldrich
Potassium gold(III) chloride, 98%
Sigma-Aldrich
Potassium gold(III) chloride, 99.995% trace metals basis
Sigma-Aldrich
Gold(III) chloride solution, 99.99% trace metals basis, 30 wt. % in dilute HCl
Sigma-Aldrich
Sodium tetrachloroaurate(III) dihydrate, 99%