Merck
  • Garcina cambogia leaf and seawater for tannase production by marine Aspergillus awamori BTMFW032 under slurry state fermentation.
Garcina cambogia leaf and seawater for tannase production by marine Aspergillus awamori BTMFW032 under slurry state fermentation.
Natural product communications (2012-02-09)
S P Beena, Soorej M Basheer, Sarita G Bhat, M Chandrasekaran
ABSTRACT
Garcinia gummi-gutta (syn. G. cambogia, G. quaesita), known to have medicinal properties, was evaluated as a substrate and inducer for tannase production by a marine Aspergillus awamori BTMFW032, under slurry state fermentation using Czapekdox-minimal medium and sea water as the cultivation medium. Among the various natural tannin substrates evaluated, Garcinia leaf supported maximal tannase production. The cultivation conditions and components of the cultivation medium were optimized employing response surface methodology. The experimental results were fitted to a second-order polynomial model at a 92.2% level of significance (p < 0.0001). The maximal tannase activity was obtained in a slurry state medium containing 26.6%, w/v, Garcinia leaf, supplemented with 0.1% tannic acid as inducer. The optimum values of pH, temperature and inoculum concentration obtained were 5.0, 40 degrees C and 3%, respectively. A Box-Behnken model study of the fermentation conditions was carried out, and the best production of tannase was registered at 40 degrees C without agitation. Optimization strategy employing response surface methodology led to nearly 3-fold increase in the enzyme production from 26.2 U/mL obtained in unoptimized medium to 75.2 Units/mL in Box Behnken design, within 18 h of fermentation. It was observed that sea water could support maximal tannase production by A. awamori compared with other media suggesting that the sea water salts could have played an inducer role in expression of tannase encoding genes. To the best of our knowledge, this is the first report on production of tannase, an industrially important enzyme, utilizing Garcinia leaf as substrate under slurry state fermentation by marine A. awamori and sea water as the cultivation medium.
Sigma-Aldrich
Tannase from Aspergillus ficuum, powder, white, ≥150 U/g