Merck
  • Adsorption of allura red dye by cross-linked chitosan from shrimp waste.
Adsorption of allura red dye by cross-linked chitosan from shrimp waste.
Water science and technology : a journal of the International Association on Water Pollution Research (2012-01-27)
Reyna G Sánchez-Duarte, Dalia I Sánchez-Machado, Jaime López-Cervantes, Ma A Correa-Murrieta
ABSTRACT
The present study was designed to evaluate the chitosan, which has been obtained by deacetylation of chitin, as a biosorbent. The chitin was isolated from fermented shrimp waste by an important local industrial food biopolymer. The aim of this work was the characterization of chitosan and preparation of cross-linked chitosan- tripolyphosphate (chitosan-TPP) beads for the removal of allura red food dye from aqueous solutions. Conditions of batch adsorption such as pH, time and adsorbent dose were examined. The effectiveness of cross-linked chitosan beads for dye removal was found to be higher for pH 2 (98%, percentage of dye removal) and tends to decrease at pHs of 3 to 11 (up to 49%). The values of percentage removal show that the adsorption capacity increases with time of contact and dosage of chitosan-TPP, but red dye adsorption is mainly influenced by pH level. The cross-linked chitosan-TPP beads can significantly adsorb allura red monoazo dye from aqueous solutions even at acidic pHs unlike raw chitosan beads that tend to dissolve in acidic solutions. Consequently, this modified chitosan has characteristics that allow minimization of environmental pollution and widening the valorization of shrimp waste.
Sigma-Aldrich
Allura Red AC, Dye content 80 %
Supelco
Allura Red AC, analytical standard