- Linking pharmacology to clinical reports: cyclobenzaprine and its possible association with serotonin syndrome.
Linking pharmacology to clinical reports: cyclobenzaprine and its possible association with serotonin syndrome.
Clinical pharmacology and therapeutics (2011-10-07)
J Mestres, S A Seifert, T I Oprea
PMID21975349
ABSTRACT
The link between cyclobenzaprine (Flexeril) administration and serotonin syndrome (SS) is subject to debate. Establishing such a connection is difficult because of the limited number of case reports available and the almost complete ignorance of its preclinical pharmacology. In this context, evidence is provided here that cyclobenzaprine blocks the serotonin and norepinephrine transporters and binds to another set of five serotonin receptors. SS should be considered when indicative signs occur in the context of cyclobenzaprine use.