Merck
  • Solvatochromic and preferential solvation studies on schiff base 1,4-bis(((2-methylthio)phenylimino)methyl) benzene in binary liquid mixtures.
Solvatochromic and preferential solvation studies on schiff base 1,4-bis(((2-methylthio)phenylimino)methyl) benzene in binary liquid mixtures.
Journal of fluorescence (2010-05-11)
G Suganthi, S Sivakolunthu, V Ramakrishnan
ABSTRACT
The solvatochromic behavior of the 1,4-bis(((2-methylthio) phenylimino)methyl) benzene [BMTPMB] in single solvents and binary mixtures were investigated. Fluorescence spectra show the dual emission due to twisted intramolecular charge transfer (TICT) state. The preferential solvation parameters: local mole fraction, X (2) (L) , solvation index δ(s2), exchange constant K(12) were calculated for the binary mixtures, ACN+MEOH, DMSO+CCl(4) and CCl(4)+1,2 DCE. The dipole moment ratios between ground and excited states were deduced using the solvatochromic shifts of absorption and fluorescence spectra as a function of dielectric constant (ε), refractive index (n) and it was found to be 1.25.
Product Description
Supelco
cis-1,2-Dichloroethene, analytical standard
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol