Merck
  • Urinary BTEX, MTBE and naphthalene as biomarkers to gain environmental exposure profiles of the general population.
Urinary BTEX, MTBE and naphthalene as biomarkers to gain environmental exposure profiles of the general population.
The Science of the total environment (2010-04-27)
Silvia Fustinoni, Federica Rossella, Laura Campo, Rosa Mercadante, Pier Alberto Bertazzi
ABSTRACT
The aim of this work was to evaluate urinary benzene, toluene, ethylbenzene, m+p-xylene, o-xylene (BTEX), methyl tert-butyl ether (MTBE), ethyl tert-butyl ether (ETBE), and naphthalene (NAP) as biomarkers of exposure to environmental pollutants. Personal air and urine samples from 108 subjects belonging to the Italian general population were compared. Urinary profiles were obtained by headspace gas chromatography-mass spectrometry. BTEX, MTBE, ETBE and NAP median airborne exposures during a 5-h sampling were 4.0, 25.3, 3.8, 9.3, 3.4, 3.4, <0.8, and 3.4 microg/m(3), respectively. Meanwhile, median urinary levels, as geometric means of three determinations were: 122, 397, 74, 127, 43, 49, <15, and 46 ng/L, respectively. Urinary benzene and toluene concentrations were 4.6- and 1.2-fold higher in smokers than in non-smokers. For most chemicals, significant positive correlations between airborne exposure (log-transformed) and the corresponding biological marker (log-transformed) were found, with Pearson's r values for correlation, ranging from 0.228 to 0.396. Multiple linear regression analysis showed that the urinary level of these chemicals was influenced by personal airborne exposure, urinary creatinine, and urinary cotinine, with R(2) 0.733 for benzene. Urinary chemicals are useful biomarkers of environmental exposure. Given the ease of rapidly obtaining urine samples, they represent a non-invasive alternative to blood chemical analysis. The possibility of obtaining urinary exposure profiles makes this method an appealing tool for environmental epidemiology.
Sigma-Aldrich
tert-Butyl ethyl ether, purum, ≥97.0% (GC)
Sigma-Aldrich
tert-Butyl ethyl ether, 99%
Supelco
tert-Butyl ethyl ether, analytical standard