Merck
  • Determination of banned Sudan dyes in food samples by molecularly imprinted solid phase extraction-high performance liquid chromatography.
Determination of banned Sudan dyes in food samples by molecularly imprinted solid phase extraction-high performance liquid chromatography.
Journal of separation science (2009-09-02)
Claudio Baggiani, Laura Anfossi, Patrizia Baravalle, Cristina Giovannoli, Gianfranco Giraudi, Claudia Barolo, Guido Viscardi
ABSTRACT
A method for molecularly imprinted SPE of banned Sudan azo-dyes from food samples was investigated. The molecularly imprinted polymer was obtained by suspension polymerization using 1-(4-chlorophenyl)azonaphthalen-2-ol as the mimic template. The molecular recognition properties of imprinted beads were evaluated for use as a SPE sorbent, in order to develop a selective extraction protocol for the Sudan class of dyes. The optimized extraction protocol resulted in a reliable molecularly imprinted SPE (MISPE) method suitable for HPLC analysis. It was selective for the main analyte, Sudan I, and the related azo-dyes Sudan II, III, IV, Sudan Red B, and Sudan Red 7B, while the permitted azo-dyes Allura Red AC, Neococcin, and Sunset Yellow FCF were not extracted. The method was tested for Sudan I, II, III, and IV in five different food samples (hot chilli pepper, hot chilli tomato sauce, sausage, tomato sauce, and hard boiled egg yolk) at three concentration levels (15, 100, and 300 microg/g). It demonstrated itself to be insensitive to the presence of different complex matrices, precise, accurate, and with good recovery rates (85-101%). The LOD and LOQ were satisfactory for most analytical determinations.
Sigma-Aldrich
Sudan II, Dye content 90 %
Supelco
Sudan II, analytical standard