Merck
Light-activation of gene function in mammalian cells via ribozymes.
Chemical communications (Cambridge, England) (2009-03-14)
Douglas D Young, R Aaron Garner, Jeffrey A Yoder, Alexander Deiters
ABSTRACT
A ribozyme based gene control element enabled the spatio-temporal regulation of gene function in mammalian cell culture with light.
Sigma-Aldrich
Toyocamycin, ≥98% (HPLC), from Streptomyces rimosus