Merck
  • Characterization of a Dehalobacter coculture that dechlorinates 1,2-dichloroethane to ethene and identification of the putative reductive dehalogenase gene.
Characterization of a Dehalobacter coculture that dechlorinates 1,2-dichloroethane to ethene and identification of the putative reductive dehalogenase gene.
Applied and environmental microbiology (2009-03-10)
Ariel Grostern, Elizabeth A Edwards
ABSTRACT
Dehalobacter and "Dehalococcoides" spp. were previously shown to be involved in the biotransformation of 1,1,2-trichloroethane (1,1,2-TCA) and 1,2-dichloroethane (1,2-DCA) to ethene in a mixed anaerobic enrichment culture. Here we report the further enrichment and characterization of a Dehalobacter sp. from this mixed culture in coculture with an Acetobacterium sp. Through a series of serial transfers and dilutions with acetate, H(2), and 1,2-DCA, a stable coculture of Acetobacterium and Dehalobacter spp. was obtained, where Dehalobacter grew during dechlorination. The isolated Acetobacterium strain did not dechlorinate 1,2-DCA. Quantitative PCR with specific primers showed that Dehalobacter cells did not grow in the absence of a chlorinated electron acceptor and that the growth yield with 1,2-DCA was 6.9 (+/-0.7) x 10(7) 16S rRNA gene copies/mumol 1,2-DCA degraded. PCR with degenerate primers targeting reductive dehalogenase genes detected three distinct Dehalobacter/Desulfitobacterium-type sequences in the mixed-parent culture, but only one of these was present in the 1,2-DCA-H(2) coculture. Reverse transcriptase PCR revealed the transcription of this dehalogenase gene specifically during the dechlorination of 1,2-DCA. The 1,2-DCA-H(2) coculture could dechlorinate 1,2-DCA but not 1,1,2-TCA, nor could it dechlorinate chlorinated ethenes. As a collective, the genus Dehalobacter has been show to dechlorinate many diverse compounds, but individual species seem to each have a narrow substrate range.
Product Description
Supelco
1,1,2-Trichloroethane, analytical standard