Merck
  • Nonionic surfactant-capped gold nanoparticles for selective enrichment of aminothiols prior to CE with UV absorption detection.
Nonionic surfactant-capped gold nanoparticles for selective enrichment of aminothiols prior to CE with UV absorption detection.
Electrophoresis (2009-02-11)
Mu-De Li, Tian-Lu Cheng, Wei-Lung Tseng
ABSTRACT
In this study, we describe the use of Tween 20-capped gold nanoparticles (AuNPs) as selective probes for the extraction of aminothiols from an aqueous solution. Tween 20 molecules noncovalently attached to the surface of AuNPs to form Tween 20-AuNPs were used for the selective extraction of aminothiols through the formation of Au-S bonds. After extraction and centrifugation, the aminothiols were detached from the surface of the AuNPs by adding DTT in a high concentration. We used this probe in combination with CE and UV absorption detection. On-line concentration and separation of the released aminothiols were performed by using 1.6% v/v poly(diallyldimethylammonium chloride) as an additive in CE. Under optimal extraction and stacking conditions, the LOD at a S/N of 3 were 28, 554, and 456 nM for glutathione (GSH), cysteine (Cys), and homocysteine (HCys), respectively. In comparison with the normal injection without the extraction procedure, approximately 2280-, 998-, and 904-fold improvements in the sensitivity were observed for GSH, Cys, and HCys, respectively. We have validated the application of our method on the basis of the analysis of GSH and HCys in human urine samples. It is believed that this approach has significant potential to be extended to clinical diagnosis.
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0% (RT), BioUltra