Merck
  • A small component of the endoplasmic reticulum is required for store-operated Ca2+ channel activation in liver cells: evidence from studies using TRPV1 and taurodeoxycholic acid.
A small component of the endoplasmic reticulum is required for store-operated Ca2+ channel activation in liver cells: evidence from studies using TRPV1 and taurodeoxycholic acid.
The Biochemical journal (2008-11-15)
Joel Castro, Edoardo C Aromataris, Grigori Y Rychkov, Greg J Barritt
ABSTRACT
The question of whether the activation of SOCs (store-operated Ca(2+) channels) requires the whole or part of the ER (endoplasmic reticulum) has not been fully resolved. The role of a putative sub-compartment of the ER in SOC activation in liver cells was investigated using ectopically expressed TRPV1 (transient receptor potential vanilloid 1), a non-selective cation channel, and TDCA (taurodeoxycholic acid), an activator of SOCs, to release Ca(2+) from different regions of the ER. TRPV1 was expressed in the ER and in the plasma membrane. The amount of Ca(2+) released from the ER by a TRPV1 agonist, measured using fura-2, was the same as that released by a SERCA (sarcoplasmic/endoplasmic reticulum Ca(2+)-ATPase) inhibitor, indicating that TRPV1 agonist-sensitive stores substantially overlap with SERCA inhibitor-sensitive stores. In contrast with SERCA inhibitors, TRPV1 agonists did not activate store-operated Ca(2+) entry. These findings were confirmed by patch-clamp recording. Using FFP-18, it was shown that SERCA inhibitors release Ca(2+) from the ER located closer to the plasma membrane than the region from which TRPV1 agonists release Ca(2+). In contrast with SERCA inhibitors, TRPV1 agonists did not induce a redistribution of STIM1 (stromal interaction molecule 1). TDCA caused the release of Ca(2+) from the ER, which was detected by FFP-18 but not by fura-2, and a redistribution of STIM1 to puncta similar to that caused by SERCA inhibitors. It is concluded that in liver cells, Ca(2+) release from a small component of the ER located near the plasma membrane is required to induce STIM1 redistribution and SOC activation.
Sigma-Aldrich
2,5-Di-tert-butylhydroquinone, 99%