Merck
  • Effects of iron type in Fenton reaction on mineralization and biodegradability enhancement of hazardous organic compounds.
Effects of iron type in Fenton reaction on mineralization and biodegradability enhancement of hazardous organic compounds.
Journal of hazardous materials (2008-05-27)
Eakalak Khan, Wanpen Wirojanagud, Nawarat Sermsai
ABSTRACT
The mineralization and biodegradability increase and their combination of two traditional and two relatively new organic contaminants by Fenton reagents with three different types of iron, Fe(2+), Fe(3+), and Fe(0) were investigated. The traditional contaminants examined were trichloroethene (TCE) and 2,4-dichlorophenol (2,4-DCP) while 1,4-dioxane (1,4-D) and 1,2,3-trichloropropane (TCP) were studied for the relatively new contaminants. The mineralization and biodegradability were represented by dissolved organic carbon (DOC) reduction and the ratio of biodegradable dissolved organic carbon and DOC, respectively. For all four contaminants, Fenton reagent using Fe(2+) was more effective in the DOC reduction than Fenton reagents using Fe(3+) and Fe(0) in most cases. The types of Fe that provided maximum biodegradability increase were not the same for all four compounds, Fe(3+) for TCE, Fe(0) for 2,4-DCP, Fe(2+) for 1,4-D, and Fe(3+) for TCP. When the combination of DOC elimination and biodegradability increase (least refractory fraction) was considered, Fe(2+) was the best choice except for 2,4-DCP which was susceptible to Fe(0) catalyzed Fenton reagent the most. The least refractory fractions remaining after 120 min of reaction were 20-25% for TCE, 2,4-DCP, and TCP and 30-40% for 1,4-D. The iron type in Fenton reaction also affected the type of mineralization kinetics of TCE, 2,4-DCP, and TCP as well as the types of degradation by-products of these contaminants. Some of the by-products found, such as isopropanol and propionic aldehyde, which were produced from Fe(0) catalyzed Fenton degradation of TCP, have not been previously reported.
Product Description
Supelco
1,2,3-Trichloropropane, analytical standard
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Available to ship on 09 January 2025
Details...
€83.60
500 g
1 Available to ship on 09 January 2025
Details...
€280.00
Supelco
1,2,3-Trichloropropane solution, certified reference material, 200 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
1 Available to ship on 09 January 2025
Details...
€83.60
500 g
1 Available to ship on 09 January 2025
Details...
€280.00