Merck
Synthesis of N-alkoxyphthalimides under ultrasound irradiation.
Ultrasonics sonochemistry (2007-09-04)
Shu-Xiang Wang, Xin-Wei Li, Ji-Tai Li
ABSTRACT
The alkylation of N-hydroxyphthalimide with alkyl halides in the presence of potassium carbonate results N-alkoxyphthalimides in 64-99% yields in DMSO under ultrasound irradiation. Compared with conventional methods, the main advantages of the present procedure are milder conditions, shorter reaction time and higher yields.
Product Description
Supelco
2-Bromoacetophenone, for GC derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
2-Bromoacetophenone, 98%