Merck
  • Immobilized cofactor derivatives for kinetic-based enzyme capture strategies: direct coupling of NAD(P)+.
Immobilized cofactor derivatives for kinetic-based enzyme capture strategies: direct coupling of NAD(P)+.
Analytical biochemistry (2004-11-24)
Laura Oakey, Patricia Mulcahy
ABSTRACT
This study reevaluates the potential for direct coupling of NAD(P)(+) to a carboxylate-terminating spacer arm using carbodiimide-promoted coupling in an attempt to develop a greatly simplified synthetic method for cofactor immobilization that would support the more widespread adoption of kinetic-based enzyme capture (KBEC) strategies for protein purification applications and protein-detecting arrays/proteomic studies. Direct coupling of NAD(+) to epoxy (1,4-butanediol diglycidyl ether)-activated Sepharose is also described. Depending on the synthetic method used, the position of attachment of cofactor is concluded to be primarily through the pyrophosphate or ribosyl hydroxyl groups. Total substitution levels varied from 0.5 to 2 micromol/g wet weight with 28-67% accessibility. Model bioaffinity chromatographic studies employing KBEC strategies are reported for bovine heart L-lactate dehydrogenase, yeast alcohol dehydrogenase, l-phenylalanine dehydrogenase from Sporosarcina, glutamate dehydrogenase (GDH) from Candida utilis, and GDH from bovine liver. The NAD(+) derivative prepared using epoxy-activated Sepharose shows most potential for further development based on total substitution levels, the apparent absence of nonbiospecific interference, reversible biospecific adsorption of some of the test enzymes using soluble KBEC/stripping ligand tactics, and the relative simplicity of the synthetic method.
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, technical grade, 60%
Sigma-Aldrich
1,4-Butanediol diglycidyl ether, ≥95%