Merck
  • Huntingtin S421 phosphorylation increases kinesin and dynein engagement on early endosomes and lysosomes.
Huntingtin S421 phosphorylation increases kinesin and dynein engagement on early endosomes and lysosomes.
Biophysical journal (2023-02-12)
Emily N P Prowse, Abdullah R Chaudhary, David Sharon, Adam G Hendricks
ABSTRACT
Huntingtin (HTT) is a scaffolding protein that recruits motor proteins to vesicular cargoes, enabling it to regulate kinesin-1, dynein, and myosin-VI-dependent transport. To maintain the native stoichiometry of HTT with its interacting partners, we used CRISPR/Cas9 to induce a phosphomimetic mutation of the endogenous HTT at S421 (HTT-S421D). Using single-particle tracking, optical tweezers, and immunofluorescence, we examined the effects of this mutation on the motility of early endosomes and lysosomes. In HTT-S421D cells, lysosomes exhibit longer displacements and higher processive fractions compared with wild-type (HTT-WT) cells. Kinesins and dyneins exert greater forces on early endosomes and lysosomes in cells expressing HTT-S421D. In addition, endosomes bind to microtubules faster and are more resistant to detachment under load. The recruitment of kinesins and dyneins to microtubules is enhanced in HTT-S421D cells. In contrast, overexpression of HTT had variable effects on the processivity, displacement, and directional bias of both early endosomes and lysosomes. These data indicate that phosphorylation of the endogenous HTT causes early endosomes and lysosomes to move longer distances and more processively by recruiting and activating both kinesin-1 and dynein.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-KIF16B antibody produced in mouse, clone 2B5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG - Atto 488 antibody produced in goat, whole molecule, 1 mg/mL protein