Merck
  • The mitochondrial ATP-dependent potassium channel (mitoKATP) controls skeletal muscle structure and function.
The mitochondrial ATP-dependent potassium channel (mitoKATP) controls skeletal muscle structure and function.
Cell death & disease (2024-01-18)
Giulia Di Marco, Gaia Gherardi, Agnese De Mario, Ilaria Piazza, Martina Baraldo, Andrea Mattarei, Bert Blaauw, Rosario Rizzuto, Diego De Stefani, Cristina Mammucari
ABSTRACT
MitoKATP is a channel of the inner mitochondrial membrane that controls mitochondrial K+ influx according to ATP availability. Recently, the genes encoding the pore-forming (MITOK) and the regulatory ATP-sensitive (MITOSUR) subunits of mitoKATP were identified, allowing the genetic manipulation of the channel. Here, we analyzed the role of mitoKATP in determining skeletal muscle structure and activity. Mitok-/- muscles were characterized by mitochondrial cristae remodeling and defective oxidative metabolism, with consequent impairment of exercise performance and altered response to damaging muscle contractions. On the other hand, constitutive mitochondrial K+ influx by MITOK overexpression in the skeletal muscle triggered overt mitochondrial dysfunction and energy default, increased protein polyubiquitination, aberrant autophagy flux, and induction of a stress response program. MITOK overexpressing muscles were therefore severely atrophic. Thus, the proper modulation of mitoKATP activity is required for the maintenance of skeletal muscle homeostasis and function.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-CCDC51 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution