Merck
  • DRP1 interacts directly with BAX to induce its activation and apoptosis.
DRP1 interacts directly with BAX to induce its activation and apoptosis.
The EMBO journal (2022-01-14)
Andreas Jenner, Aida Peña-Blanco, Raquel Salvador-Gallego, Begoña Ugarte-Uribe, Cristiana Zollo, Tariq Ganief, Jan Bierlmeier, Markus Mund, Jason E Lee, Jonas Ries, Dirk Schwarzer, Boris Macek, Ana J Garcia-Saez
ABSTRACT
The apoptotic executioner protein BAX and the dynamin-like protein DRP1 co-localize at mitochondria during apoptosis to mediate mitochondrial permeabilization and fragmentation. However, the molecular basis and functional consequences of this interplay remain unknown. Here, we show that BAX and DRP1 physically interact, and that this interaction is enhanced during apoptosis. Complex formation between BAX and DRP1 occurs exclusively in the membrane environment and requires the BAX N-terminal region, but also involves several other BAX surfaces. Furthermore, the association between BAX and DRP1 enhances the membrane activity of both proteins. Forced dimerization of BAX and DRP1 triggers their activation and translocation to mitochondria, where they induce mitochondrial remodeling and permeabilization to cause apoptosis even in the absence of apoptotic triggers. Based on this, we propose that DRP1 can promote apoptosis by acting as noncanonical direct activator of BAX through physical contacts with its N-terminal region.
Sigma-Aldrich
Monoclonal Anti-6X His IgG, CF680 antibody produced in mouse, ~2 mg/mL, clone 33D10.D2.G8, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-MUL1 antibody produced in rabbit
SKU
Pack Size
Availability
Price
Quantity
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)