Merck
  • PDGF-AA activates AKT and ERK signaling for testicular interstitial Leydig cell growth via primary cilia.
PDGF-AA activates AKT and ERK signaling for testicular interstitial Leydig cell growth via primary cilia.
Journal of cellular biochemistry (2022-10-29)
Yung-Chieh Tsai, Tian-Ni Kuo, Yu-Ying Chao, Pei-Rong Lee, Ruei-Ci Lin, Xiao-Yi Xiao, Bu-Miin Huang, Chia-Yih Wang
ABSTRACT
Testes control the development of male reproductive system. The testicular interstitial Leydig cells (Leydig cells) synthesize testosterone for promoting spermatogenesis and secondary sexual characteristics. Type A platelet-derived growth factor (PDGF-AA) is one of the most important growth factors in regulating Leydig cell growth and function. Knockout of PDGF-AA or its congenital receptor PDGFR-α leads to poor testicular development caused by reducing Leydig cell numbers, supporting PDGF-AA/PDGFR-α signaling regulates Leydig cell development. Primary cilium is a cellular antenna that functions as an integrative platform to transduce extracellular signaling for proper development and differentiation. Several receptors including PDGFR-α are observed on primary cilia for initiating signaling cascades in distinct cell types. Here we showed that PDGF-AA/PDGFR-α signaling promoted Leydig cells growth, migration, and invasion via primary cilia. Upon PDGF-AA treatment, AKT and ERK signaling were activated to regulate these cellular events. Interestingly, active AKT and ERK were detected around the base of primary cilia. Depletion of ciliary genes (IFT88 and CEP164) alleviated PDGF-AA-activated AKT and ERK, thus reducing Leydig cell growth, migration, and invasion. Thus, our study not only reveals the function of PDGF-AA/PDGFR-α signaling in maintaining testicular physiology but also uncovers the role of primary cilium and downstream signaling in regulating Leydig cell development.
Sigma-Aldrich
Akt Inhibitor IV, The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid